Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI3D8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270407JF549

Catalog No. N270407J549


Add to cart

Description

Description

CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

CEACAM5/CD66e
OTI3D8
Immunohistochemistry
Affinity Purified, Conjugated
CEACAM5
Recombinant protein fragment corresponding to amino acids 35-680 of human CEACAM5/CD66e (NP_004354) produced in SF9 cell.
Immunogen affinity purified
Apoptosis, Cancer, Cellular Markers, Immunology
Primary
1048.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit