Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Mouse anti-Human, PE/Atto594, Clone: 4H6-20, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254629PEATT594

Catalog No. N254629P594


Add to cart

Description

Description

CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CEACAM5/CD66e
4H6-20
Flow Cytometry
Conjugated, Purified
CEACAM5
Recombinant human CEA
Protein A or G purified
Apoptosis, Cancer, Cellular Markers, Immunology
Primary
1048.0
Human
Flow Cytometry
PE/ATTO 594
PBS with 0.05% Sodium Azide
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
This antibody recognizes human CEACAM5,6 & 8.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit