Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Mouse anti-Human, PE-Cyanine7, Clone: C66/1292, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254412PECY7

Catalog No. N254412PEC7


Add to cart

Description

Description

CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CEACAM5/CD66e
C66/1292
Flow Cytometry
Conjugated, Purified
CEACAM5
Recombinant full-length human CEA protein
Protein A or G purified
Apoptosis, Cancer, Cellular Markers, Immunology
Primary
1048.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
IgG2b
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit