Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5/CD66e Rabbit anti-Human, Clone: 76, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289786100UL
Description
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for ELISA.Specifications
CEACAM5/CD66e | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
76 | |
ELISA 1:5000-1:10000 | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
Recombinant Human CEACAM5/CD66e protein | |
100 μg | |
Apoptosis, Cancer, Cellular Markers, Immunology | |
1048 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title