Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM5/CD66e Rabbit anti-Human, Clone: 76, Novus Biologicals™

Rabbit Monoclonal Antibody

$321.00 - $493.00

Specifications

Antigen CEACAM5/CD66e
Clone 76
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB021043
View Documents
Novus Biologicals
NBP289786100UL
100 μg
Each for $493.00
Add to cart
 
NB021044
View Documents
Novus Biologicals
NBP28978650UL
50 μg
Each for $321.00
Add to cart
 
Description

Description

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for ELISA.
Specifications

Specifications

CEACAM5/CD66e
ELISA 1:5000-1:10000
Monoclonal
Purified
RUO
Human
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Recombinant Human CEACAM5/CD66e protein
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
76
ELISA
Unconjugated
Rabbit
Apoptosis, Cancer, Cellular Markers, Immunology
0.2 um filtered solution in PBS
1048
IgG
Protein A purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit