Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CEACAM5/CD66e Rabbit anti-Human, Janelia Fluor 646, Clone: Arcitumomab, Novus Biologicals
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP252673JF646
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
CEACAM5/CD66e | |
Arcitumomab | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Purified | |
CEACAM5 | |
Carcinoembryonic antigen (CEA) describes a set of highly related glycoproteins involved in cell adhesion normally produced in gastrointestinal tissue during fetal development, but only at very low levels in adults. | |
Protein A or G purified | |
Apoptosis, Cancer, Cellular Markers, Immunology | |
Primary | |
1048 | |
Human, Rat |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
Rabbit | |
IgG Kappa | |
0.1mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Recognises human carcinoembryonic antigen. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title