Learn More
Novus Biologicals™ CEACAM6/CD66c Antibody Pair [Biotin]
Polyclonal/Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2793035PLATES
 View more versions of this product
Catalog No. NB009646
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human CEACAM6/CD66c.
Specifications
Specifications
Biotin
Antibody Pairs
15.6 pg/mL
4680
RUO
CEACAM6/CD66c
Cancer
Human
CEACAM6
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1000 pg/mL is recommended.
Sandwich ELISA
This solid phase sandwich ELISA utilizes an antibody specific for Human CEACAM6/CD66c.
Sandwich ELISA
  • Detection Antibody: 0.4 mg/mL of biotinylated rabbit anti-CEACAM6/CD66c polyclonal antibody.
  • Capture Antibody: 1 mg/mL of mouse anti-CEACAM6/CD66c monoclonal antibody.
  • Standard: Each vial contains 88 ng of recombinant CEACAM6/CD66c.
  • Storage is content dependent.
    Rabbit/Mouse
    5 Plates
    15.6-1000 pg/mL
    carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen