Learn More
Novus Biologicals™ CEACAM6/CD66c Antibody Pair [HRP]
Rabbit Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2792885PLATES
 View more versions of this product
Catalog No. NB009616
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CEACAM6/CD66c.
Specifications
Specifications
Sandwich ELISA
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human CEACAM6/CD66c.
9.34 pg/mL
4680
RUO
CEACAM6/CD66c
Cancer
Human
CEACAM6
1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 600 pg/mL is recommended.
HRP
Antibody Pairs
Sandwich ELISA
  • Standard: Each vial contains 40 ng of recombinant Human CEACAM6/CD66c.
  • Capture Antibody: 0.4 mg/mL of rabbit anti-Human CEACAM6/CD66c monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.15 mg/mL of rabbit anti-Human CEACAM6/CD66c monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Storage is content dependent.
    Rabbit
    5 Plates
    9.34-600 pg/mL
    carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen