Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CEACAM6/CD66c Mouse anti-Human, Allophycocyanin/Cy7, Clone: 1H7-4B, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254626APCCY7

Catalog No. N254626APC7


Add to cart

Description

Description

CEACAM6/CD66c Monoclonal antibody specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CEACAM6/CD66c
1H7-4B
Flow Cytometry
Conjugated, Purified
CEACAM6
Recombinant human CEACAM6 extracellular domain.
Protein A or G purified
Cancer
Primary
4680.0
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit