Learn More
Alfa Aesar™ Cecropin A (1-8)-Melittin (1-18)Amide
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66814MCR
 View more versions of this product
Catalog No. AAJ66814MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2794.50
1mg
Lys-Trp-Lys-Leu-Phe-Lys-Lys-Ile-Gly-Ile-Gly-Ala-Val-Leu-Lys-Val-Leu-Thr-Thr-Gly-Leu-Pro-Ala-Leu-Ile-Ser-NH2
-30° to -10°C
HPLC
Lyophilized powder
RUO
Cecropin A (1-8)-Melittin (1-18) amide