Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cedarwood Oil, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
35 lb.
Packaging:
Amber Glass Bottle
Steel Pail
68990-83-0
68990-83-0
Quantity 35 lb.
CAS Max % 1
Packaging Steel Pail
Refractive Index 1.510 -1.520
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.