Learn More
Thermo Scientific CEDIA™ Therapeutic Drug Monitoring (TDM) Assays Available on GSA/VA Contract for Federal Government customers only.
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays.
Manufacturer:  Thermo Scientific 100018
 View more versions of this product
Catalog No. 23-278-236
  Check Availability
Add to cart
Specifications
Description
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.
  • Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
  • Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range
Description & Specifications
Specifications
Tobramycin II Assay
13, 11mL
2° to 8°C
Tobramycin
Plasma
Reagent