Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays.
Supplier: Thermo Scientific™ 100087
This item is not returnable.
View return policy
Description
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.
- Wide dynamic measuring range of analytes as low as 0.15ng/mL for Digoxin
- Extended linearity for analytes like Cyclosprine at 25ng/mL to 2000ng/mL range
Specifications
2°C to 8°C | |
NAPA | |
Reagent |
NAPA Assay | |
Plasma, Serum | |
17 x 17 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction