Learn More
Abnova™ CEECAM1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00051148T01
Catalog No. 89-939-176
Add to cart
Description
Description
Specifications
Specifications
Western Blot
CEECAM1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 56.98kDa
Transient overexpression cell lysate was tested with anti-CEECAM1 antibody (Mfr. No. H00051148-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_016174.3
pCMV-CEECAM1 full-length
100 μL
Human