Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefadroxil, MilliporeSigma™ Supelco™

Cefadroxil is a broad spectrum cephalosporin antibiotic, widely used in veterinery medicine for the oral treatment of infectious diseases.

$53.40 - $167.70

Specifications

CAS 50370-12-2
Grade Analytical Standard
Linear Formula C16H17N3O5 S
Physical Form Powder or Crystals
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4362
View Documents
MilliporeSigma Supelco
C70201G
1 g
Each for $53.40
Add to cart
 
11-101-4363
View Documents
MilliporeSigma Supelco
C70205G
5 g
Each for $167.70
Add to cart
 
Description

Description

Featured Industries: Pharmaceutical (small molecule)
Specifications

Specifications

50370-12-2
Analytical Standard
C16H17N3O5 S
Powder or Crystals
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit