Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefazolin, USP, 95-103%, Spectrum™

Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail

C2522, 25953-19-9, C14H14N8O4S3

Spectrum™ Cefazolin, USP is used as a cephalosporin antibiotic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Water 0.02
Quantity 5 kg
CAS 25953-19-9
CAS Max % 1
Percent Purity 95 to 103%