Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefdinir, 95%

Click to view available options
Quantity:
1g
5g
10g
91832-40-5
C14H13N5O5S2
395.408
MFCD00865030
RTXOFQZKPXMALH-GHXIOONMSA-N
6915944
CHEBI:3485
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O
91832-40-5
C14H13N5O5S2
395.408
MFCD00865030
RTXOFQZKPXMALH-GHXIOONMSA-N
6915944
CHEBI:3485
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-hydroxyiminoacetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NO)C3=CSC(=N3)N)SC1)C(=O)O
  • A broad spectrum antibiotic targeting both gram positive and gram negative pathogens
Solubility Information Soluble in water
Formula Weight 395.41
Physical Form Powder
Percent Purity 95%
Color White to Yellow
Melting Point 170°C
Quantity 1g
Chemical Name or Material Cefdinir

RTECSNumber : XI0367250

storageNote1 : 2° to 8°C