Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefepime hydrochloride monohydrate

Click to view available options
Quantity:
1g
5g
123171-59-5
C19H29Cl2N6O6S2+
572.497
MFCD01687679
LRAJHPGSGBRUJN-OMIVUECESA-O
129893984
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
123171-59-5
C19H29Cl2N6O6S2+
572.497
MFCD01687679
LRAJHPGSGBRUJN-OMIVUECESA-O
129893984
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-[(1-methylpyrrolidin-1-ium-1-yl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
C[N+]1(CCCC1)CC2=C(N3C(C(C3=O)NC(=O)C(=NOC)C4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
Applications
Cefepime hydrochloride monohydrate is used as a broad spectrum antibiotic targeting a wide variety of naturally antibiotic resistant Gram-positive and Gram-negative bacteria. It is resistant to hydrolysis by common plasmid and/or chromosomally-mediated β-lactamases.

Solubility
Soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Solubility Information Soluble in DMSO
Formula Weight 571.5
Physical Form Powder
Color White to Yellow
Melting Point 150°C
Quantity 1g
Sensitivity Light Sensitive
Chemical Name or Material Cefepime hydrochloride monohydrate

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : 2° to 8°C