Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefixime trihydrate

Click to view available options
Quantity:
100mg
1g
79350-37-1
C16H15N5O7S2
453.444
MFCD03788802
OKBVVJOGVLARMR-QSWIMTSFSA-N
5362065
CHEBI:472657
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NOCC(=O)O)C3=CSC(=N3)N)SC1)C(=O)O
79350-37-1
C16H15N5O7S2
453.444
MFCD03788802
OKBVVJOGVLARMR-QSWIMTSFSA-N
5362065
CHEBI:472657
(6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(carboxymethoxyimino)acetyl]amino]-3-ethenyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C=CC1=C(N2C(C(C2=O)NC(=O)C(=NOCC(=O)O)C3=CSC(=N3)N)SC1)C(=O)O

A broad spectrum third generation cephalosporin

Solubility Information Soluble in water at 55.1μg/mL
Formula Weight 507.49
Physical Form Powder
Color Yellow
Melting Point 218°C to 225°C
Quantity 100mg
Sensitivity Air & Light Sensitive
Chemical Name or Material Cefixime trihydrate

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0367200

storageNote1 : 2° to 8°C