Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefmetazole

Click to view available options
Quantity:
1g
5g
56796-20-4
C15H17N7O5S3
471.525
MFCD00865068
SNBUBQHDYVFSQF-HIFRSBDPSA-N
42008
CHEBI:3489
(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)O
56796-20-4
C15H17N7O5S3
471.525
MFCD00865068
SNBUBQHDYVFSQF-HIFRSBDPSA-N
42008
CHEBI:3489
(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)O

A broad spectrum second generation cephalosporin antibiotic

Solubility Information Soluble in water
Formula Weight 471.53
Physical Form Powder
Color White
Quantity 1g
Chemical Name or Material Cefmetazole

RTECSNumber : XI0372000

storageNote1 : 2° to 8°C