Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefmetazole sodium, 921μg/mg

Click to view available options
Quantity:
1g
5g
56796-39-5
C15H16N7NaO5S3
493.507
MFCD00214260
BITQGIOJQWZUPL-PBCQUBLHSA-M
23666711
CHEBI:3490
sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+]
56796-39-5
C15H16N7NaO5S3
493.507
MFCD00214260
BITQGIOJQWZUPL-PBCQUBLHSA-M
23666711
CHEBI:3490
sodium;(6R,7S)-7-[[2-(cyanomethylsulfanyl)acetyl]amino]-7-methoxy-3-[(1-methyltetrazol-5-yl)sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CN1C(=NN=N1)SCC2=C(N3C(C(C3=O)(NC(=O)CSCC#N)OC)SC2)C(=O)[O-].[Na+]
  • Cefmetazole is used to study the effect of expression, binding, and inhibition of penicillin-binding proteins (with the exception of PBP2) on bacterial cell wall mucopeptide synthesis
  • It is also used to study protein mediated antibiotic tr
Solubility Information Soluble in water
Formula Weight 493.52
Physical Form Powder
Color White
Quantity 5g
Sensitivity Hygroscopic
Chemical Name or Material Cefmetazole sodium, 921μg/mg

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0372200

storageNote1 : 2° to 8°C