Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefotiam dihydrochloride

Click to view available options
Quantity:
50mg
100mg
66309-69-1
C18H27Cl2N9O5S3
616.552
MFCD09878605
KHUCCKUOJMXYKH-BUWDDJPCSA-N
70590037
(6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl
66309-69-1
C18H27Cl2N9O5S3
616.552
MFCD09878605
KHUCCKUOJMXYKH-BUWDDJPCSA-N
70590037
(6R,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)acetyl]amino]-3-[[1-[2-(dimethylamino)ethyl]tetrazol-5-yl]sulfanylmethyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate;dihydrochloride
CN(C)CCN1C(=NN=N1)SCC2=C(N3C(C(C3=O)NC(=O)CC4=CSC(=N4)N)SC2)C(=O)O.O.Cl.Cl

A broad spectrum antibiotic

Solubility Information Soluble in water
Formula Weight 562.09
Physical Form Powder
Boiling Point 94°C
Color White
Quantity 100mg
Chemical Name or Material Cefotiam dihydrochloride

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : XI0367000

storageNote1 : 2° to 8°C