Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefoxitin sodium, 92.7-97%

Click to view available options
Quantity:
250mg
1g
33564-30-6
C16H16N3NaO7S2
449.428
MFCD00079042
GNWUOVJNSFPWDD-XMZRARIVSA-M
23667300
CHEBI:3501
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
33564-30-6
C16H16N3NaO7S2
449.428
MFCD00079042
GNWUOVJNSFPWDD-XMZRARIVSA-M
23667300
CHEBI:3501
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]

A broad spectrum antibiotic

Merck Index 14,1936
Solubility Information Soluble in water or methanol
Formula Weight 449.43
Physical Form Powder
Percent Purity 92.7 to 97%
Boiling Point 843°C
Color White
Melting Point >160°C
Quantity 1g
Chemical Name or Material Cefoxitin sodium

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XI0330500

storageNote1 : 2° to 8°C