Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefoxitin sodium salt, 94%, Acros Organics™

Click to view available options
Quantity:
1g
5g
33564-30-6
C16H16N3NaO7S2
449.428
GNWUOVJNSFPWDD-XMZRARIVSA-M
23667300
CHEBI:3501
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
33564-30-6
C16H16N3NaO7S2
449.428
GNWUOVJNSFPWDD-XMZRARIVSA-M
23667300
CHEBI:3501
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
Infrared Spectrum Authentic
Formula Weight 449.43
Percent Purity 94%
Specific Rotation +206° to +214° (20°C, 589nm) (c= 1, methanol)
Water 1% max.
Quantity 5g
Chemical Name or Material Cefoxitin sodium salt

GHS H Statement May cause an allergic skin reaction.

GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. If skin irritation or rash occurs: Get medical advice/attention.

Warning