Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefpirome sulfate, 95%

Click to view available options
Quantity:
1g
5g
98753-19-6
C22H24N6O9S3
612.647
MFCD11864966
RKTNPKZEPLCLSF-PXPMWPIZSA-N
134129654
(6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]
98753-19-6
C22H24N6O9S3
612.647
MFCD11864966
RKTNPKZEPLCLSF-PXPMWPIZSA-N
134129654
(6S,7R)-7-[[2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-3-(6,7-dihydro-5H-cyclopenta[b]pyridin-1-ium-1-ylmethyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrogen sulfate
CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC5=C4CCC5)C(=O)O.OS(=O)(=O)[O-]

A fourth generation cephalosporin antibiotic

Solubility Information Soluble in DMSO
Formula Weight 612.66
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 198°C to 202°C
Quantity 1g
Chemical Name or Material Cefpirome sulfate

RTECSNumber : UW8970000

storageNote1 : 2° to 8°C