Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefradine

Click to view available options
Quantity:
1g
5g
25g
38821-53-3
C16H19N3O4S
349.405
MFCD00865048
RDLPVSKMFDYCOR-UEKVPHQBSA-N
38103
CHEBI:3547
(6R,7R)-7-[[(2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CC1=C(N2C(C(C2=O)NC(=O)C(C3=CCC=CC3)N)SC1)C(=O)O
38821-53-3
C16H19N3O4S
349.405
MFCD00865048
RDLPVSKMFDYCOR-UEKVPHQBSA-N
38103
CHEBI:3547
(6R,7R)-7-[[(2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
CC1=C(N2C(C(C2=O)NC(=O)C(C3=CCC=CC3)N)SC1)C(=O)O
  • A semi-synthetic cephalosporin antibiotic that is used to study the effect of expression, binding, and inhibition of PBP3 and other penicillin-binding proteins (PBPs) on bacterial cell wall mucopeptide synthesis
Merck Index 14,1986
Solubility Information Soluble in water
Formula Weight 349.4
Physical Form Powder
Boiling Point 898°C
Color White to Yellow
Melting Point 140°C to 142°C
Quantity 25g
Chemical Name or Material Cefradine

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0336000

storageNote1 : 2° to 8°C

Recommended Storage : 2° to 8°C