Learn More
Cefsulodin sodium salt, 50 mg/ml in distilled water, sterile-filtered, Thermo Scientific Chemicals
Description
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.
Solubility
Soluble in water.
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Specifications
Type | Cefsulodin Sodium Salt |
Physical Form | Liquid |
Solubility Information | Soluble in water. |
Quantity | 1 mL |
Chemical Name or Material | Cefsulodin sodium salt, 50mg/mL in distilled water, sterile-filtered |
Safety and Handling
Hazard Category | H317-H334-H335 |
Hazard Statement | GHS H Statement H334-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. |
Precautionary Statement | P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c |
EINECSNumber | 257-692-4 |
RTECSNumber | UU1785000 |
TSCA | Yes |
storageNote1 | -30° to -10°C |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.