Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefsulodin sodium salt, 50mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
52152-93-9
C22H21N4NaO8S2+
556.54
MFCD07793338
GTZPOHRNWUTXNB-DWBVFMGKSA-O
124203950
(6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]
52152-93-9
C22H21N4NaO8S2+
556.54
MFCD07793338
GTZPOHRNWUTXNB-DWBVFMGKSA-O
124203950
(6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium
C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na]
Applications
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.

Solubility
Soluble in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Cefsulodin sodium salt, 50mg/mL in distilled water, sterile-filtered

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : UU1785000

TSCA : TSCA

storageNote1 : -30° to -10°C