Learn More
Cefsulodin sodium salt hydrate, 97%, Thermo Scientific Chemicals
Description
Cefsulodin is most commonly used in CIN agar to select for Yersinia microorganisms. The compound displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP) crosslinking of peptidoglycan resulting in inhibition of the final transpeptidation step. Through the inability for Cefsulodin sodium salt hydrate to inhibit cefsulodin-resistant mutants of Pseudomonas aeruginosa PAO4089 growth displayed that Cefsulodin sodium salt hydrate may compete with PBP3 in addition to PBP1A and PBP1B.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Cefsulodin sodium is a third-generation cephalosporin antibiotic with a narrow range of effectiveness
• Cefsulodin competitively inhibits penicillin binding protein, preventing cell wall synthesis and promoting lysis
Application
• Cefsulodin is mainly effective against Yersinia microorganisms, Pseudomonas aeruginosa, and Staphylococcus aureus
• Cefsulodin is useful for drug resistance studies involving susceptible microbes and for studies related to drug mechanisms
• It is appropriate for selection experiments and other cell culture applications
Specifications
Type | Cefsulodin Sodium Salt |
Physical Form | Powder |
Melting Point | 175°C |
Molecular Formula | C22H21N4NaO8S2+ |
Merck Index | 14,1945 |
Solubility Information | Freely soluble in water |
InChI Key | GTZPOHRNWUTXNB-DWBVFMGKSA-O |
SMILES | C1C(=C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)S(=O)(=O)O)C(=O)O)C[N+]4=CC=C(C=C4)C(=O)N.[Na] |
IUPAC Name | (6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;sodium |
Molecular Weight (g/mol) | 556.54 |
Show More |
Safety and Handling
Hazard Category | H315-H317-H319-H334-H335 |
Hazard Statement | GHS H Statement H334-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
Precautionary Statement | P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c |
EINECSNumber | 257-692-4 |
RTECSNumber | UU1785000 |
TSCA | Yes |
storageNote1 | -30° to -10°C |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.