Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ceftibuten, 95%

Click to view available options
Quantity:
10mg
50mg
100mg
97519-39-6
C15H14N4O6S2
410.419
MFCD00864918
UNJFKXSSGBWRBZ-BJCIPQKHSA-N
5282242
CHEBI:3510
(6R,7R)-7-[[(Z)-2-(2-amino-1,3-thiazol-4-yl)-4-carboxybut-2-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C1C=C(N2C(S1)C(C2=O)NC(=O)C(=CCC(=O)O)C3=CSC(=N3)N)C(=O)O
97519-39-6
C15H14N4O6S2
410.419
MFCD00864918
UNJFKXSSGBWRBZ-BJCIPQKHSA-N
5282242
CHEBI:3510
(6R,7R)-7-[[(Z)-2-(2-amino-1,3-thiazol-4-yl)-4-carboxybut-2-enoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
C1C=C(N2C(S1)C(C2=O)NC(=O)C(=CCC(=O)O)C3=CSC(=N3)N)C(=O)O

A broad spectrum antibiotic

Applications
Ceftibuten is used to treat acute bacterial exacerbations of chronic bronchitis (ABECB), acute bacterial otitis media, pharyngitis, andtonsilitis. It is also indicated for pneumonia, infections of the urinary tract, enteritis, and gastroenteritis.

Solubility
Soluble in aqueous solutions. Also soluble in DMSO

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Solubility Information Soluble in aqueous solutions; Also soluble in DMSO
Formula Weight 410.42
Physical Form Powder
Percent Purity 95%
Boiling Point 966°C
Color Brown to White
Quantity 50mg
Chemical Name or Material Ceftibuten

RTECSNumber : XI0367220

storageNote1 : 2° to 8°C