Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftizoxime sodium, 95%

Click to view available options
Quantity:
50mg
68401-82-1
C13H12N5NaO5S2
405.379
MFCD01682036
ADLFUPFRVXCDMO-YKEGMJKBSA-M
122130206
sodium;(7R)-7-[[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=CCS3)C(=O)[O-].[Na+]
68401-82-1
C13H12N5NaO5S2
405.379
MFCD01682036
ADLFUPFRVXCDMO-YKEGMJKBSA-M
122130206
sodium;(7R)-7-[[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=CCS3)C(=O)[O-].[Na+]
  • A broad spectrum antibiotic targeting a wide variety of gram positive and gram negative bacteria
Solubility Information Soluble in water
Formula Weight 405.38
Physical Form Powder
Percent Purity 95%
Color White to Yellow
Melting Point 227°C
Quantity 50mg
Chemical Name or Material Ceftizoxime sodium

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : XI0368000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold