Learn More
Alkali Scientific Cefuroxime, Sodium Salt, 5 Grams
SDP
Supplier:  Alkali Scientific EFU002CP
Encompass
Cefuroxime Sodium Salt, 5g is a broad-spectrum antibiotic used in laboratory research for antimicrobial studies. It is commonly employed to evaluate bacterial susceptibility to antibiotics and to test the efficacy of various antimicrobial agents. Cefuroxime is a second-generation cephalosporin antibiotic that inhibits bacterial cell wall synthesis, leading to bacterial death. This 5g vial is ideal for preparing larger volumes of solution for in vitro testing and antimicrobial research applications. It is often used in pharmaceutical research and microbiological investigations, particularly in studies related to bacterial infections and resistance mechanisms.
Catalog No. 50-006-59883