Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Cefuroxime sodium salt, 95%

Click to view available options
Quantity:
1g
5g
56238-63-2
C16H15N4NaO8S
446.366
MFCD09878727
URDOHUPGIOGTKV-LNJPBTIASA-M
133687825
sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+]
56238-63-2
C16H15N4NaO8S
446.366
MFCD09878727
URDOHUPGIOGTKV-LNJPBTIASA-M
133687825
sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+]
Merck Index 14,1954
Solubility Information Freely soluble in water
Formula Weight 446.37
Physical Form Powder
Percent Purity 95%
Color White to Yellow
Melting Point 240°C to 245°C
Quantity 1g
Chemical Name or Material Cefuroxime sodium salt

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P285-P261-P280-P342+P311-P363-P501a
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : XI0330000

storageNote1 : 2° to 8°C