Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CELA1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548907
Description
Acts upon elastin.Specifications
CELA1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UNI1 | |
CELA1 | |
KLH conjugated synthetic peptide between 51-79 amino acids from the N-terminal region of human CELA1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
CELA1 | |
Chymotrypsin-like elastase family member 1, Elastase-1, Pancreatic elastase 1, CELA1, ELA1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1990 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction