Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CELA3B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548629

Catalog No. PA548629

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Efficient protease with alanine specificity but only little elastolytic activity.
Specifications

Specifications

CELA3B
Polyclonal
PBS with 0.09% sodium azide
P08861
Chymotrypsin-like elastase family member 3B, Elastase IIIB, Elastase-3B, Protease E, CELA3B, ELA3B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23436
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CELA3B
Liquid
CELA3B
KLH conjugated synthetic peptide between 70-98 amino acids from the N-terminal region of human CELA3B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit