Learn More
Celestron™ Telescope Accessories Products for Science Education
Manufacturer:  Celestron™ 93319
 View more versions of this product
Catalog No. S68082
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Image Erecting Eyepiece (IEP)
·    Device turns astronomical telescopes into terrestrial viewing telescopes
·    Corrects inverted image produced by an astronomical telescope
·    Turn it “right side up” to enable terrestrial viewing
·    For use with 1 1/4 in. eyepieces
Specifications
Specifications
Omni 12 mm Eyepiece
2 years
0.49 in.
12 mm