Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celite™, for analysis, high purity analytical grade, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
61790-53-2
O2Si
60.083
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Infusorial earth,Diatomite,Diatomaceous earth
24261
CHEBI:30563
dioxosilane
O=[Si]=O
61790-53-2
O2Si
60.083
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Infusorial earth,Diatomite,Diatomaceous earth
24261
CHEBI:30563
dioxosilane
O=[Si]=O
Filter aid
Color Gray-Pink to White
pH 5 to 8 (25°C, 10% aq. suspension)
Quantity 10g
Acid Soluble Iron acid soluble iron: <5ppm
Calcium Oxide 0.1%
Arsenic (As) 10ppm max.
Conductivity 15μS/cm (15μmho/cm) Max.
Fe2O3 0.3%
Lead (Pb) 10ppm max.
Loss on Drying 0.5% max.
Loss on Ignition 2% max.
Nonsiliceous Substances Nonsiliceous substances < 50mg
Packaging Glass bottle
Permeability 0.15 to 0.30 DARCY
Silicon Dioxide 98.7%
Wet Density centrifuged wet density < 0.25g/mL (centrifuged)
Formula Weight 60.08
Physical Form Powder
Grade Analysis
Chemical Name or Material Celite™
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.