Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Celite(R), 501, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Fiber Drum
Poly Pail
68855-54-9
68855-54-9
Quantity 2.5 kg
CAS Max % 1
Packaging Poly Pail
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.