Learn More
Celite|r 535, Thermo Scientific Chemicals
$82.00 - $182.00
Specifications
Melting Point | >450°C |
---|---|
CAS | 68855-54-9 |
Molecular Formula | SiO{2} |
Synonym | Diatomaceous Earth;Filter aid, flux calcined |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAH377580B
|
Thermo Scientific Chemicals
H377580B |
1000 g |
Each for $82.00
Order in Bulk
Request bulk or custom formats |
|
|||||
AAH377580I
|
Thermo Scientific Chemicals
H377580I |
5000 g |
Each for $182.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
>450°C | |
68855-54-9 | |
SiO{2} | |
Diatomaceous Earth;Filter aid, flux calcined |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.