Learn More
Mirus™ Cell Culture Antibiotic-Antimycotic Solution (100X)
Effective for preventing bacterial and fungal contamination of tissue culture cells
Manufacturer:  Mirus™ MIR 5970
Catalog No. MIR5970
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Light yellow solution
  • High Potency, Easy to use, Broad spectrum
  • Composed of Penicillin (10,000 units/mL), Streptomycin (10,000 μg/mL), and Amphotericin B (25μg/mL)
  • Molecular formula: C16H18N2O4S (Penicillin); C21H39N7O12 (Streptomycin); C47H73NO17 (Amphotericin B)
  • Effective against gram-positive and gram-negative bacteria, yeast and other fungi
  • Recommended use concentration for  Antibiotic-Antimycotic Solution in culture media is 1X (10mL/L)
  • Individual cell lines may react differently to this product
  • Testing multiple concentrations on target cell line to evaluate tolerance recommended
  • Observe cells daily for signs of toxicity such as loss of confluence, changes in morphology, or detachment
  • Decrease Antibiotic-Antimycotic concentration in media if cytotoxicity occurs