Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Cell Culture Dishes, Tissue Culture Treated

For cell based assays and microscopic analysis. Eppendorf™ Cell Culture Dishes, Tissue Culture Treated helps in direct access to the cells.

Supplier:  Eppendorf™ 0030700112

 View more versions of this product

Catalog No. 13-690-079

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Safety in handling during transportation and while working due to corrugated handling ring
  • SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
  • Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
  • Easy differentiation of lid and dish bottom prevents unintended removal of the lid
  • Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
  • Available in three formats: 35, 60 and 100mm
Specifications

Specifications

Transparent
Polystyrene
Flat
Transparent
Polystyrene
Sterile
Cell Culture Dish
Lid
40.8 mm
Closure Included
Not Autoclavable
8.1 mL
9.5 sq. cm
Round
Tissue Culture
2 to 3 mL
10.8 mm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit