Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ Cell Culture Dishes, Tissue Culture Treated
For cell based assays and microscopic analysis. Eppendorf™ Cell Culture Dishes, Tissue Culture Treated helps in direct access to the cells.
Supplier: Eppendorf™ 0030702115
Description
- Safety in handling during transportation and while working due to corrugated handling ring
- SplashProtect ring inside of the dish lid traps liquid and prevents spills during transportation or incubation
- Pronounced rim on dish lid and exact fitting of lid and bottom ensure sturdy, secure stacking
- Easy differentiation of lid and dish bottom prevents unintended removal of the lid
- Packaging combines tool-free opening, resealable top and side for tape-free closing, and an easy solution to shrink the package for space-conscious storage
- Available in three formats: 35, 60 and 100mm
Specifications
Transparent | |
Polystyrene | |
Flat | |
Transparent | |
Polystyrene | |
Sterile | |
Cell Culture Dish | |
Lid | |
93.8 mm |
Closure Included | |
Not Autoclavable | |
99.2 mL | |
56.8 sq. cm | |
Round | |
Tissue Culture | |
8 to 10 mL | |
19.4 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title