Learn More
Abbott Laboratories™ CELL-DYN Emerald 22
Hematology analyzer for smaller clinical laboratories seeking better productivity with small spaces.
Supplier:  Abbott Laboratories™ 09H5901
Catalog No. 09H5901A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Optical Flow Cytometry technology
  • Electrical impedance
  • Absorption spectrophotometry
  • Electronic valves
  • Cyanide-free lyse reagent
  • LCD color touch screen
  • RS232 and TCP/IP LIS interface
  • USB ports
  • Up to 45 samples per hour
  • Up to 300,000 records on external USB thumb drive
  • Operator-defined patient limits for high and panic values
  • System-defined limits for reportable range and analytical measurement range
  • Suspect parameter flags caused by interfering substances or sample abnormalities
  • Suspect parameter flags generated when WBC data indicates possible presence of an abnormal population
Specifications
Specifications
17 μL
20
24.2 lb.
9.8 in.
13.8 in.
45/hr
13.8 in.