Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abbott Laboratories™ CELL-DYN Emerald 22 Autoloader
Automated hematology analyzer delivering 5-part differential for low- to mid-volume laboratories.
Supplier: Abbott Laboratories™ 04R1301
This item is not returnable.
View return policy
Description
- Open and closed sampling
- Reagents: 2 and Cleaner
- Auto rerun: Yes
- Load up Capacity: 50 tubes with 5 per rack
- Maintenance Requirements: 2 minute bleach clean cycle weekly, manual syringe lubrication biannually
Specifications
Open: 21 μL; Closed: 21 μL | |
16.1 in. | |
40/hr | |
19.7 in. |
16.9 in. | |
20 | |
55 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title