Learn More
BioVision IncSupplier Diversity Partner FractionPREP™ Cell Fractionation Kit: Isolate Four Subcellular Fractions: Cytosol, Nucleus, Membrane/Particulate and Cytoskeletal from a single mammalian sample within 2 hours without ultracentrifugation. 50 Fractionations. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K27050
This FractionPREP™ Cell Fractionation system is designed to provide reproducible extraction of four subcellular protein fractions (Cytosol, nucleus, membrane/particulate, and cytoskeletal fractions) from a single mammalian sample. The method is fast and simple, needing only 2 hours and no ultracentrifugation involved. All four protein fractions obtained are suitable for many downstream applications such as 1-D or 2-D gel, enzyme activity assays, gel shift assay, and Western blotting. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9663