Learn More
Enzo Life SciencesSupplier Diversity Partner Cell lysis buffer 5, (100 ml) (100 ml)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADI801631
Formulation: Liquid. 20mM TRIS, pH 7.4, containing 500mM sodium chloride, 10mM EDTA, 1% Triton X-100, 20mM sodium fluoride, 20mM beta-glycerophosphate. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C
Catalog No. 50-658-076