Learn More
Cell Biolabs Inc
CELL MIGR ASSAY 24W 8UM COLOR
Manufacturer: Cell Biolabs Inc CBA1005
The CytoSelect 24-Well Cell Migration Assay Kit contains twelve 8 um polycarbonate membrane inserts in a 24-well plate, which migratory cells are able to pass through. Quantify cells using the provided cell stain solution. 5x12 inserts.