Learn More
BioVision IncSupplier Diversity Partner EZCell™ Cell Migration/Chemotaxis Assay Kit (96-well, 8 µm): Highly Sensitive and Quantitative Fluorometric Assay to measure Cell Migration/Chemotaxis using a Boyden Chamber. 8 µm, 100 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K906100
Cell migration is a process by which cells move from one location to another. Cell motility is observed in unicellular organisms, and is essential for the development and maintenance of multicellular organisms. Cells often migrate in response to specific external stimuli, including chemical & mechanical signals. Errors during this process have serious consequences, including intellectual disability, vascular disease, tumor formation and metastasis. BioVision’s Cell Migration/Chemotaxis Assay Kit utilizes a Boyden chamber, where the cells migrate through a semipermeable membrane under different stimuli. Cell migration can be analyzed directly by reading fluorescence (Ex/Em = 530/590 nm) in a plate reader. Our assay is easy to use, sensitive and adaptable to high-throughput systems. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9973