Learn More
BioVision IncSupplier Diversity Partner Quick Cell Proliferation colorimetric Assay Kit Plus: Easy & Sensitive WST based Colorimetric Assay to Measure Cell Proliferation in response to growth factors, cytokines, mitogens and nutrients etc. Kit Size: 500 & 2500 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K302500
Quick Cell Proliferation colorimetric Assay Kit Plus: The Quick Cell Proliferation Assay Kit II provides by far the easiest and most sensitive means for quantifying cell proliferation and viability. The amount of the dye generated by activity of dehydrogenase is directly proportional to the number of living cells. The dye produced by viable cells can be quantified by microtiter plate reader by measuring the absorbance of the dye solution at 440 nm. The assay can be used for measurement of cell proliferation in response to growth factors, cytokines, mitogens, and nutrients, etc. It can also be used for the analysis of cytotoxic compounds like anticancer drugs and many other toxic agents and pharmaceutical compounds. The new method is so simple, just add-and-read, requiring no washing, no harvesting, and no solubilization steps. It is faster, stable, and more sensitive than MTT, XTT, MTS based assays. The assay correlates well with the [3H]-thymidine incorporation assay.
Catalog No. 50-111-9677