Learn More
Cell Size and Diffusion Lab
A quick, easy and safe visual demonstration of cell size and diffusion.
Manufacturer: Neo/Sci Corporation 201015
Includes:
- Agar
- Diffusion media
- Cups, gloves and rulers
- Dilute base
- Comprehensive teacher and student guides
Description
Students construct and analyze simulated cell models of various sizes using a specially formulated agar in the process, they learn how cells obtain nutrients and excrete waste. Data obtained by measuring the cell's volume and calculating its surface area is used to form conclusions about the size limitations imposed by diffusion.
Order Info
Class size: 40 students.
Specifications
Why Cells Aren'T Big Refill Kit | |
40 students | |
Agar, diffusion media, cups, gloves and rulers, dilute base, comprehensive teacher and student guides |
30 minutes | |
Easy and safe visual demonstration of cell size & diffusion |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.